Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145221.2 | NP_660256.1 | 17 | 1699 | ||
GenBank mRNA | AK030445.1 | - | 17 | 1578 | ||
AK033566.1 | - | 17 | 1670 | |||
AK045438.1 | - | 17 | 1635 | |||
AK049307.1 | - | 17 | 1686 | |||
AK146356.1 | - | 17 | 1670 | |||
AK173169.1 | - | 2 | 150 | |||
AY113705.1 | - | 17 | 1664 | |||
BC019708.1 | - | 9 | 977 | |||
BC063751.1 | - | 17 | 1711 | 6511401 |