Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271531.1 | NP_001258460.1 | 3 | 511 | ||
NM_001271532.1 | NP_001258461.1 | 2 | 378 | |||
NM_133748.2 | NP_598509.1 | 3 | 754 | |||
NM_178082.3 | NP_835183.1 | 3 | 518 | |||
GenBank mRNA | AF527631.1 | - | 3 | 488 | ||
AK040703.1 | - | 3 | 509 | |||
AK152076.1 | - | 3 | 484 | |||
AK154543.1 | - | 3 | 517 | |||
AK161256.1 | - | 3 | 522 | |||
AK165634.1 | - | 3 | 523 | |||
BC015288.1 | - | 3 | 468 | 4240711 | ||
BC023067.1 | - | 3 | 492 | 5357599 | ||
BC023874.1 | - | 2 | 410 | 5355039 | ||
BC024411.1 | - | 3 | 506 | 4971297 |