Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028431.2 | NP_082707.1 | 10, 11 | 1258 | ||
GenBank mRNA | AK013995.1 | - | 10, 11 | 1259 | ||
AK152673.1 | - | 10, 11 | 1260 | |||
AK169417.1 | - | 10, 11 | 1215 | |||
BC038321.1 | - | 9, 10 | 1129 | |||
BC051216.1 | - | 9, 10 | 1065 | |||
BC060662.1 | - | 9, 10 | 3770 | |||
BC065095.1 | - | 9, 10 | 3770 | |||
BC087943.1 | - | 10, 11 | 1270 |