Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm7sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028454.2 | NP_082730.2 | 4 | 546 | ||
GenBank mRNA | AF480070.1 | - | 4 | 420 | ||
AK014166.1 | - | 4 | 527 | |||
AK037325.1 | - | 3 | 350 | |||
AK039028.1 | - | 4 | 476 | |||
AK043687.1 | - | 3 | 729 | |||
AK134512.1 | - | 1 | 1247 | |||
AK155475.1 | - | 4 | 503 | |||
BC014769.1 | - | 4 | 538 | 4239159 |