Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032413.1 | NP_001027585.1 | 7 | 774 | ||
NM_001032414.1 | NP_001027586.1 | 6 | 704 | |||
NM_001289600.1 | NP_001276529.1 | 6 | 676 | |||
NM_001289601.1 | NP_001276530.1 | 8 | 917 | |||
NM_028460.2 | NP_082736.1 | 8 | 908 | |||
NM_152799.2 | NP_690012.2 | 8 | 873 | |||
GenBank mRNA | AF440279.1 | - | 8 | 898 | ||
AF444274.1 | - | 6 | 698 | |||
AF461685.1 | - | 5 | 631 | |||
AK014179.1 | - | 6 | 715 | |||
AK042308.1 | - | 7 | 765 | |||
AK053551.1 | - | 7 | 765 | |||
AK141082.1 | - | 671 | ||||
AK161049.1 | - | 8 | 914 | |||
BC042490.1 | - | 7 | 723 | 4953584 |