Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028480.2 | NP_082756.1 | 5, 6 | 1696 | ||
GenBank mRNA | AK145976.1 | - | 5, 6 | 1697 | ||
AK146102.1 | - | 5, 6 | 1698 | |||
BC100349.1 | - | 5, 6 | 1674 |