Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028481.1 | NP_082757.1 | 27 | 3824 | ||
GenBank mRNA | AK016519.1 | - | 27 | 3818 | ||
AK045927.1 | - | 239 | ||||
BC082598.1 | - | 27 | 3824 | 30641632 |