Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270435.1 | NP_001257364.1 | 10 | 1610 | ||
NM_001270446.1 | NP_001257375.1 | 5 | 866 | |||
NM_028506.1 | NP_082782.1 | 6 | 873 | |||
NM_029286.2 | NP_083562.1 | 10 | 1625 | |||
GenBank mRNA | AK049010.1 | - | 5 | 766 | ||
AK078362.1 | - | 10 | 1552 | |||
AK163185.1 | - | 10 | 1551 | |||
AK224940.1 | - | 6 | 802 | |||
AK224959.1 | - | 5 | 774 | |||
AK224961.1 | - | 5 | 867 | |||
BC114436.1 | - | 10 | 1552 | 40059415 |