Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027022.4 | NP_081298.1 | 1 | 238 | ||
GenBank mRNA | AK005603.1 | - | 1 | 191 | ||
AK006874.1 | - | 1 | 239 | |||
AK161303.1 | - | 1 | 232 | |||
AY162281.1 | - | 1 | 119 | |||
AY172740.1 | - | 1 | 119 | |||
AY241871.1 | - | 1 | 238 | |||
BC049747.1 | - | 1 | 231 | 6772661 |