Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290783.1 | NP_001277712.1 | 6 | 1346 | ||
NM_026137.5 | NP_080413.2 | 6 | 1380 | |||
NM_028542.1 | NP_082818.1 | 6 | 1632 | |||
GenBank mRNA | AF220146.1 | - | 5 | 902 | ||
AF353243.1 | - | 6 | 1026 | |||
AF513713.1 | - | 6 | 849 | |||
AK006837.1 | - | 5 | 868 | |||
AK017549.1 | - | 6 | 962 | |||
BC057366.1 | - | 6 | 1346 | |||
BC080853.1 | - | 6 | 1295 | |||
BC108332.1 | - | 6 | 1136 | 6493226 | ||
BC120553.1 | - | 6 | 883 | 40129476 | ||
BC125287.1 | - | 6 | 882 | 40129802 |