Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028543.3 | NP_082819.1 | 4 | 531 | ||
GenBank mRNA | AK088750.1 | - | 4 | 532 | ||
AK141711.1 | - | 4 | 532 | |||
BC056193.1 | - | 4 | 518 | 30011543 |