Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028557.2 | NP_082833.1 | 3 | 476 | ||
GenBank mRNA | AK006927.1 | - | 3 | 477 | ||
AK018931.1 | - | 3 | 471 | |||
AY038023.1 | - | 2 | 425 | |||
BC049738.1 | - | 3 | 492 | 6771508 | ||
BC061034.1 | - | 3 | 482 | 6707322 |