Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028576.2 | NP_082852.1 | 5, 6 | 516 | ||
GenBank mRNA | AK155246.1 | - | 5, 6 | 517 | ||
BC138272.1 | - | 5, 6 | 664 | 8861291 | ||
BC138273.1 | - | 5, 6 | 664 | 8861292 |