Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023712.3 | NP_076201.2 | 9 | 2065 | ||
GenBank mRNA | AF212372.1 | - | 9 | 1556 | ||
AK008721.1 | - | 10 | 1541 | |||
AK033907.1 | - | 7 | 2620 | |||
AK154353.1 | - | 9 | 1555 | |||
AK166931.1 | - | 9 | 1557 | |||
AK195969.1 | - | 3 | 373 | |||
BC002297.1 | - | 9 | 1084 | |||
BC026854.1 | - | 9 | 1526 | 4242049 | ||
BC085491.1 | - | 9 | 1509 | 6829415 |