Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023712.3 | NP_076201.2 | 7, 8 | 1831 | ||
GenBank mRNA | AF212372.1 | - | 7, 8 | 1322 | ||
AK008721.1 | - | 8, 9 | 1307 | |||
AK033907.1 | - | 5, 6 | 2386 | |||
AK154353.1 | - | 7, 8 | 1321 | |||
AK166931.1 | - | 7, 8 | 1323 | |||
AK187798.1 | - | 1, 2 | 49 | |||
AK195969.1 | - | 1, 2 | 140 | |||
AK199707.1 | - | 1, 2 | 49 | |||
AK206636.1 | - | 1, 2 | 49 | |||
BC002297.1 | - | 7, 8 | 850 | |||
BC026854.1 | - | 7, 8 | 1292 | 4242049 | ||
BC085491.1 | - | 7, 8 | 1275 | 6829415 | ||
BY384284.1 | - | 1, 2 | 138 |