Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028599.2 | NP_082875.1 | 13 | 1430 | ||
GenBank mRNA | AK010772.1 | - | 13 | 1430 | ||
AK152412.1 | - | 13 | 1431 | |||
AK163221.1 | - | 13 | 1404 | |||
AK168032.1 | - | 13 | 1424 | |||
AK168163.1 | - | 13 | 1432 | |||
AK169932.1 | - | 13 | 1428 | |||
BC049925.1 | - | 2 | 162 | |||
BC055483.1 | - | 2 | 331 | |||
BC085177.1 | - | 13 | 1460 | |||
BC117880.2 | - | 13 | 1460 | 40096010 | ||
BC117881.1 | - | 13 | 1457 | 40096014 |