Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028599.2 | NP_082875.1 | 14 | 1599 | ||
GenBank mRNA | AK010772.1 | - | 14 | 1599 | ||
AK152412.1 | - | 14 | 1600 | |||
AK163221.1 | - | 14 | 1573 | |||
AK168032.1 | - | 14 | 1593 | |||
AK168163.1 | - | 14 | 1601 | |||
AK169932.1 | - | 14 | 1597 | |||
BC049925.1 | - | 3 | 331 | |||
BC055483.1 | - | 3 | 500 | |||
BC085177.1 | - | 14 | 1629 | |||
BC117880.2 | - | 14 | 1629 | 40096010 | ||
BC117881.1 | - | 14 | 1626 | 40096014 |