Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081076.2 | NP_001074545.1 | 8 | 1493 | ||
GenBank mRNA | AY395631.1 | - | 8 | 1493 |