Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028636.2 | NP_082912.1 | 14 | 1617 | ||
GenBank mRNA | AK036765.1 | - | 653 | |||
AK049370.1 | - | 14 | 1638 | |||
AK153935.1 | - | 14 | 1638 | |||
AK163077.1 | - | 14 | 1618 | |||
AK170642.1 | - | 14 | 1647 | |||
AK171110.1 | - | 15 | 1750 | |||
AK192462.1 | - | 3 | 177 | |||
AK192594.1 | - | 3 | 177 | |||
AK197697.1 | - | 3 | 177 | |||
AK201109.1 | - | 3 | 177 | |||
AK205207.1 | - | 3 | 177 | |||
AK208918.1 | - | 3 | 177 | |||
AK210371.1 | - | 3 | 177 | |||
AK214329.1 | - | 3 | 258 | |||
AK214350.1 | - | 3 | 258 | |||
AK214526.1 | - | 4 | 358 | |||
BC016253.1 | - | 14 | 1617 | 4911979 |