Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290662.1 | NP_001277591.1 | 6, 7 | 640 | ||
NM_134471.4 | NP_608301.3 | 6, 7 | 681 | |||
GenBank mRNA | AK088047.1 | - | 6, 7 | 622 | ||
AK150222.1 | - | 6, 7 | 612 | |||
AK161309.1 | - | 6, 7 | 640 | |||
BC006841.1 | - | 6, 7 | 641 | 3598157 |