Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178628.5 | NP_848743.1 | 13 | 1837 | ||
GenBank mRNA | AK015327.1 | - | 6 | 848 | ||
AK046919.1 | - | 12 | 1567 | |||
AK050339.1 | - | 13 | 1674 | |||
AK077004.1 | - | 1 | 959 | |||
AK083918.1 | - | 703 | ||||
AK140185.1 | - | 12 | 1579 | |||
AK147311.1 | - | 13 | 5101 | |||
BC050784.1 | - | 12 | 1566 | 6595181 | ||
BC057068.1 | - | 8 | 1398 | 5689726 |