Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028705.3 | NP_082981.3 | 2 | 149 | ||
GenBank mRNA | AK017531.1 | - | 2 | 122 | ||
BC042574.1 | - | 2 | 161 | 4503584 | ||
BC145978.1 | - | 2 | 151 | 40131281 | ||
BC145980.1 | - | 2 | 151 | 40131283 |