Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028705.3 | NP_082981.3 | 24 | 2981 | ||
GenBank mRNA | AK122192.1 | - | 21 | 3937 | ||
BC022719.1 | - | 2 | 221 | |||
BC042574.1 | - | 24 | 2993 | 4503584 | ||
BC145978.1 | - | 24 | 2983 | 40131281 | ||
BC145980.1 | - | 24 | 2983 | 40131283 | ||
EF601879.1 | - | 3 | 463 |