Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025947.2 | NP_001021118.1 | 10 | 1260 | ||
NM_001276340.1 | NP_001263269.1 | 10 | 1278 | |||
NM_001276341.1 | NP_001263270.1 | 9 | 1022 | |||
NM_152816.3 | NP_690029.2 | 11 | 1299 | |||
GenBank mRNA | AB079133.1 | - | 10 | 1112 | ||
AK018195.1 | - | 11 | 1239 | |||
AK051443.1 | - | 10 | 1189 | |||
AK080871.1 | - | 10 | 1180 | |||
AK154123.1 | - | 10 | 1029 | |||
BC040777.1 | - | 9 | 992 | 4948368 | ||
BC079635.1 | - | 10 | 1109 | 6400313 | ||
BC085158.1 | - | 11 | 1204 | |||
BQ550714.1 | - | 2 | 196 | |||
CD559921.1 | - | 2 | 122 | |||
CK391623.1 | - | 2 | 47 | |||
CN525473.1 | - | 5 | 650 | |||
CO432650.1 | - | 7 | 788 | |||
DV652251.1 | - | 3 | 286 |