Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017525.1 | NP_001017525.1 | 15 | 1866 | ||
NM_028709.2 | NP_082985.2 | 17 | 3719 | |||
GenBank mRNA | AK018115.1 | - | 7 | 1180 | ||
AK039038.1 | - | 14 | 1662 | |||
AK044022.1 | - | 15 | 1757 | |||
BC059036.1 | - | 5 | 737 | |||
BC072592.1 | - | 17 | 2344 | |||
BC096535.1 | - | 17 | 3719 | |||
BC141145.1 | - | 17 | 2791 | 9055757 |