Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017525.1 | NP_001017525.1 | 4, 5 | 583 | ||
NM_028709.2 | NP_082985.2 | 6, 7 | 2436 | |||
GenBank mRNA | AK039038.1 | - | 3, 4 | 379 | ||
AK044022.1 | - | 4, 5 | 474 | |||
BC072592.1 | - | 6, 7 | 1061 | |||
BC096535.1 | - | 6, 7 | 2436 | |||
BC141145.1 | - | 6, 7 | 1508 | 9055757 |