Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028719.1 | NP_082995.1 | 13, 14 | 1474 | ||
GenBank mRNA | AK014396.1 | - | 10, 11 | 1230 | ||
AK043692.1 | - | 13, 14 | 1487 | |||
BC043087.1 | - | 13, 14 | 1474 | 6417611 | ||
BC063081.1 | - | 13, 14 | 1474 | 6417611 |