Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014568.1 | - | 8 | 1522 | ||
AK042581.1 | - | 8 | 1508 | |||
AK046761.1 | - | 8 | 1513 | |||
AK047599.1 | - | 6 | 2165 | |||
AK050948.1 | - | 8 | 1520 | |||
AK157398.1 | - | 8 | 1520 | |||
AK157956.1 | - | 9 | 1425 | |||
AK164493.1 | - | 8 | 1531 | |||
AK172043.1 | - | 8 | 1526 | |||
AK198530.1 | - | 1 | 107 | |||
AK203665.1 | - | 1 | 107 | |||
BC024877.1 | - | 8 | 1524 | |||
BC051109.1 | - | 7 | 1405 | |||
BC096599.1 | - | 7 | 1424 | |||
BC125432.1 | - | 8 | 1519 | 40129947 | ||
BC132099.1 | - | 8 | 1519 | 40130376 |