Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014568.1 | - | 7 | 1269 | ||
AK042581.1 | - | 7 | 1255 | |||
AK046761.1 | - | 7 | 1260 | |||
AK047599.1 | - | 5 | 1912 | |||
AK050948.1 | - | 7 | 1267 | |||
AK079935.1 | - | 7 | 1268 | |||
AK157398.1 | - | 7 | 1267 | |||
AK157956.1 | - | 8 | 1172 | |||
AK164493.1 | - | 7 | 1278 | |||
AK172043.1 | - | 7 | 1273 | |||
AK179132.1 | - | 1 | 25 | |||
BC024877.1 | - | 7 | 1271 | |||
BC051109.1 | - | 6 | 1152 | |||
BC096599.1 | - | 6 | 1171 | |||
BC125432.1 | - | 7 | 1266 | 40129947 | ||
BC132099.1 | - | 7 | 1266 | 40130376 |