Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271411.1 | NP_001258340.1 | 4 | 757 | ||
NM_001271412.1 | NP_001258341.1 | 4 | 687 | |||
NM_001271413.1 | NP_001258342.1 | 4 | 596 | |||
NM_001271414.1 | NP_001258343.1 | 4 | 684 | |||
NM_028728.3 | NP_083004.1 | 4 | 614 | |||
GenBank mRNA | AF361364.1 | - | 4 | 691 | ||
AK014802.2 | - | 4 | 615 | |||
AK048992.1 | - | 4 | 598 | |||
AK150635.1 | - | 4 | 666 | |||
AK151187.1 | - | 4 | 666 | |||
AK151884.1 | - | 4 | 666 | |||
AK152262.1 | - | 4 | 666 | |||
AK152499.1 | - | 4 | 666 | |||
AK152688.1 | - | 4 | 666 | |||
AK155106.1 | - | 4 | 611 | |||
AK162939.1 | - | 4 | 668 | |||
AK169918.1 | - | 4 | 611 | |||
AK170098.1 | - | 4 | 701 | |||
AK170355.1 | - | 4 | 664 | |||
AK170442.1 | - | 4 | 666 | |||
AK170545.1 | - | 4 | 552 | |||
AK170593.1 | - | 4 | 622 | |||
AK170677.1 | - | 4 | 622 | |||
AK170811.1 | - | 4 | 622 | |||
AK170883.1 | - | 4 | 665 | |||
AK171082.1 | - | 4 | 727 | |||
AK171086.1 | - | 4 | 635 | |||
AK171201.1 | - | 4 | 596 | |||
BC150153.1 | - | 4 | 629 | |||
BC150488.1 | - | 4 | 629 | 40141021 |