Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271411.1 | NP_001258340.1 | 2 | 573 | ||
NM_001271412.1 | NP_001258341.1 | 2 | 503 | |||
NM_001271413.1 | NP_001258342.1 | 2 | 412 | |||
NM_001271414.1 | NP_001258343.1 | 2 | 500 | |||
NM_028728.3 | NP_083004.1 | 2 | 430 | |||
GenBank mRNA | AF361364.1 | - | 2 | 507 | ||
AK014802.2 | - | 2 | 431 | |||
AK048992.1 | - | 2 | 414 | |||
AK150635.1 | - | 2 | 482 | |||
AK151187.1 | - | 2 | 482 | |||
AK151884.1 | - | 2 | 482 | |||
AK152262.1 | - | 2 | 482 | |||
AK152499.1 | - | 2 | 482 | |||
AK152688.1 | - | 2 | 482 | |||
AK155106.1 | - | 2 | 427 | |||
AK162939.1 | - | 2 | 484 | |||
AK169918.1 | - | 2 | 427 | |||
AK170098.1 | - | 2 | 517 | |||
AK170355.1 | - | 2 | 480 | |||
AK170442.1 | - | 2 | 482 | |||
AK170545.1 | - | 2 | 368 | |||
AK170593.1 | - | 2 | 438 | |||
AK170677.1 | - | 2 | 438 | |||
AK170811.1 | - | 2 | 438 | |||
AK170883.1 | - | 2 | 481 | |||
AK171082.1 | - | 2 | 543 | |||
AK171086.1 | - | 2 | 451 | |||
AK171201.1 | - | 2 | 412 | |||
BC150153.1 | - | 2 | 445 | |||
BC150488.1 | - | 2 | 445 | 40141021 |