Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028746.3 | NP_083022.1 | 1 | 700 | ||
GenBank mRNA | AB072352.1 | - | 1 | 683 | ||
AJ417662.1 | - | 1 | 649 | |||
AK002431.1 | - | 1 | 703 | |||
AK128901.1 | - | 1 | 700 | |||
AK143768.1 | - | 1 | 702 | |||
AK165403.1 | - | 1 | 701 | |||
BC014684.1 | - | 1 | 673 | 4236308 |