Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028749.1 | NP_083025.1 | 9, 10 | 742 | ||
GenBank mRNA | AK002734.1 | - | 9, 10 | 742 | ||
AK088859.1 | - | 9, 10 | 744 | |||
BC022734.1 | - | 9, 10 | 738 | 4506776 |