Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024258.2 | NP_077220.2 | 14 | 1869 | ||
GenBank mRNA | AK004594.2 | - | 14 | 1844 | ||
AK158778.1 | - | 3 | 3155 | |||
AK160038.1 | - | 14 | 1839 | |||
BC003278.1 | - | 14 | 1798 | 3582180 | ||
BC004577.1 | - | 2 | 381 |