Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028773.3 | NP_083049.1 | 2 | 259 | ||
GenBank mRNA | AJ306422.1 | - | 2 | 125 | ||
AK004734.2 | - | 2 | 260 | |||
AK156202.1 | - | 2 | 324 | |||
AK171017.1 | - | 2 | 257 | |||
BC064734.1 | - | 2 | 244 |