Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028780.3 | NP_083056.2 | 3, 4 | 1408 | ||
GenBank mRNA | AF269152.1 | - | 3, 4 | 1056 | ||
AK004754.2 | - | 3, 4 | 1410 | |||
AK090065.1 | - | 2, 3 | 1403 | |||
AK152476.1 | - | 3, 4 | 1188 | |||
AK152540.1 | - | 3, 4 | 1188 | |||
BC007187.1 | - | 2, 3 | 713 | |||
BC017617.1 | - | 2, 3 | 1347 | 4923337 |