Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081158.2 | NP_001074627.1 | 4 | 608 | ||
GenBank mRNA | AB093256.1 | - | 2 | 155 | ||
AK036540.1 | - | 4 | 592 | |||
AK145708.1 | - | 4 | 649 | |||
AK150045.1 | - | 4 | 567 | |||
AK154146.1 | - | 4 | 597 | |||
AK154925.1 | - | 4 | 609 | |||
AK157310.1 | - | 4 | 618 | |||
BC072573.1 | - | 4 | 563 | 5705116 |