Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081158.2 | NP_001074627.1 | 2 | 217 | ||
GenBank mRNA | AK036540.1 | - | 2 | 201 | ||
AK145708.1 | - | 2 | 257 | |||
AK150045.1 | - | 2 | 176 | |||
AK154146.1 | - | 2 | 206 | |||
AK154925.1 | - | 2 | 218 | |||
AK157310.1 | - | 2 | 227 | |||
AK210771.1 | - | 1 | 23 | |||
BC072573.1 | - | 2 | 172 | 5705116 |