Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028790.3 | NP_083066.1 | 7 | 754 | ||
GenBank mRNA | AB078618.1 | - | 7 | 751 | ||
AK004905.1 | - | 7 | 755 | |||
AK033663.1 | - | 7 | 754 | |||
AK034622.1 | - | 7 | 758 | |||
BC025852.1 | - | 7 | 707 | |||
BC132050.1 | - | 7 | 785 | 40130327 | ||
BC137996.1 | - | 7 | 785 | 8861015 |