Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpcpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042672.1 | NP_001036137.1 | 9 | 944 | ||
NM_001291050.1 | NP_001277979.1 | 8 | 817 | |||
NM_001291051.1 | NP_001277980.1 | 8 | 847 | |||
NM_001291052.1 | NP_001277981.1 | 8 | 865 | |||
NM_027096.2 | NP_081372.1 | 7 | 741 | |||
NM_028802.3 | NP_083078.3 | 9 | 941 | |||
GenBank mRNA | AK009563.1 | - | 2 | 160 | ||
AK030645.1 | - | 9 | 895 | |||
AK031658.1 | - | 8 | 820 | |||
AK049491.1 | - | 1 | 30 | |||
AK050318.1 | - | 9 | 856 | |||
AK141374.1 | - | 9 | 830 | |||
AK163419.1 | - | 8 | 787 | |||
AK165952.1 | - | 9 | 906 | |||
AK166454.1 | - | 8 | 830 | |||
BC006887.1 | - | 8 | 789 | |||
BC033408.1 | - | 7 | 671 | 5357683 |