Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpcpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042672.1 | NP_001036137.1 | 19 | 1953 | ||
NM_001291050.1 | NP_001277979.1 | 18 | 1826 | |||
NM_001291051.1 | NP_001277980.1 | 18 | 1856 | |||
NM_001291052.1 | NP_001277981.1 | 18 | 1874 | |||
NM_027096.2 | NP_081372.1 | 17 | 1750 | |||
NM_028802.3 | NP_083078.3 | 19 | 1950 | |||
GenBank mRNA | AK009563.1 | - | 12 | 1169 | ||
AK030645.1 | - | 19 | 1904 | |||
AK031658.1 | - | 18 | 1829 | |||
AK049491.1 | - | 11 | 1039 | |||
AK050318.1 | - | 19 | 1865 | |||
AK051728.1 | - | 11 | 1013 | |||
AK122510.1 | - | 6 | 2605 | |||
AK141374.1 | - | 19 | 1840 | |||
AK163419.1 | - | 18 | 1797 | |||
AK165952.1 | - | 19 | 1917 | |||
AK166293.1 | - | 10 | 884 | |||
BC006887.1 | - | 18 | 1798 | |||
BC033408.1 | - | 17 | 1680 | 5357683 |