Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpcpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042672.1 | NP_001036137.1 | 17 | 1623 | ||
NM_001291050.1 | NP_001277979.1 | 16 | 1496 | |||
NM_001291051.1 | NP_001277980.1 | 16 | 1526 | |||
NM_001291052.1 | NP_001277981.1 | 16 | 1544 | |||
NM_027096.2 | NP_081372.1 | 15 | 1420 | |||
NM_028802.3 | NP_083078.3 | 17 | 1620 | |||
GenBank mRNA | AK009563.1 | - | 10 | 839 | ||
AK030645.1 | - | 17 | 1574 | |||
AK031658.1 | - | 16 | 1499 | |||
AK049491.1 | - | 9 | 709 | |||
AK050318.1 | - | 17 | 1535 | |||
AK051728.1 | - | 9 | 683 | |||
AK122510.1 | - | 4 | 2275 | |||
AK141374.1 | - | 17 | 1510 | |||
AK163419.1 | - | 16 | 1467 | |||
AK166293.1 | - | 8 | 554 | |||
AK166454.1 | - | 16 | 1509 | |||
BC006887.1 | - | 16 | 1468 | |||
BC033408.1 | - | 15 | 1350 | 5357683 |