Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028805.2 | NP_083081.2 | 18, 19 | 2077 | ||
GenBank mRNA | AK010364.1 | - | 18, 19 | 2008 | ||
AK031630.1 | - | 18, 19 | 1989 | |||
AK034103.1 | - | 18, 19 | 1997 | |||
AK037858.1 | - | 2309 | ||||
AK049345.1 | - | 18, 19 | 1991 | |||
AK054173.1 | - | 18, 19 | 2049 | |||
AK087973.1 | - | 18, 19 | 1990 | |||
AK136322.1 | - | 18, 19 | 1998 | |||
AK184639.1 | - | 3 | 197 | |||
BC024500.1 | - | 9, 10 | 884 | |||
BC045200.1 | - | 17, 18 | 1988 | 5322489 |