Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028805.2 | NP_083081.2 | 19, 20 | 2214 | ||
GenBank mRNA | AK010364.1 | - | 19, 20 | 2142 | ||
AK031630.1 | - | 19, 20 | 2126 | |||
AK034103.1 | - | 19, 20 | 2134 | |||
AK037858.1 | - | 2446 | ||||
AK049345.1 | - | 19, 20 | 2128 | |||
AK054173.1 | - | 19, 20 | 2186 | |||
AK087973.1 | - | 19, 20 | 2127 | |||
AK136322.1 | - | 19, 20 | 2135 | |||
BC024500.1 | - | 10, 11 | 1021 | |||
BC045200.1 | - | 18, 19 | 2125 | 5322489 |