Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028805.2 | NP_083081.2 | 16 | 1907 | ||
GenBank mRNA | AK010364.1 | - | 16 | 1838 | ||
AK031630.1 | - | 16 | 1819 | |||
AK034103.1 | - | 16 | 1827 | |||
AK037858.1 | - | 2065 | ||||
AK049345.1 | - | 16 | 1821 | |||
AK054173.1 | - | 16 | 1879 | |||
AK087973.1 | - | 16 | 1820 | |||
AK136322.1 | - | 16 | 1828 | |||
AK184639.1 | - | 1 | 26 | |||
BC024500.1 | - | 7 | 714 | |||
BC045200.1 | - | 15 | 1818 | 5322489 |