Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253812.1 | NP_001240741.1 | 12, 13 | 1436 | ||
NM_028811.3 | NP_083087.1 | 12, 13 | 1379 | |||
GenBank mRNA | AK012072.1 | - | 12, 13 | 1317 | ||
AK088457.1 | - | 12, 13 | 1400 | |||
AK150058.1 | - | 12, 13 | 1317 | |||
BC026461.1 | - | 10, 11 | 1031 | |||
BC037470.1 | - | 6, 7 | 733 | |||
BC048822.1 | - | 7, 8 | 884 | |||
BC057453.1 | - | 12, 13 | 1319 | 3969625 |