Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4enif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166547.1 | NP_001160019.1 | 7, 8 | 1180 | ||
NM_001166548.1 | NP_001160020.1 | 7, 8 | 1184 | |||
NM_001166549.1 | NP_001160021.1 | 7, 8 | 1288 | |||
NM_023743.2 | NP_076232.2 | 7, 8 | 1195 | |||
GenBank mRNA | AB031388.1 | - | 7, 8 | 1016 | ||
AK041598.1 | - | 7, 8 | 1146 | |||
AK088425.1 | - | 2, 3 | 282 | |||
AK128990.1 | - | 7, 8 | 1286 | |||
AK162367.1 | - | 1, 2 | 127 | |||
AK165678.1 | - | 7, 8 | 1183 | |||
AK205705.1 | - | 2, 3 | 361 | |||
BC033410.1 | - | 7, 8 | 1030 | 5359290 |