Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gale | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178389.3 | NP_848476.1 | 7, 8 | 960 | ||
GenBank mRNA | AK009083.1 | - | 8, 9 | 828 | ||
AK146184.1 | - | 8, 9 | 851 | |||
BC027438.1 | - | 7, 8 | 960 | 4224021 |