Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021535.4 | NP_067510.3 | 10 | 1292 | ||
GenBank mRNA | AB044414.1 | - | 10 | 1221 | ||
AK003493.1 | - | 10 | 1279 | |||
AK019174.1 | - | 10 | 1251 | |||
AK145982.1 | - | 10 | 1291 | |||
AK159635.1 | - | 10 | 1279 | |||
AK168813.1 | - | 10 | 1286 | |||
AK212788.1 | - | 1 | 97 | |||
BC020034.1 | - | 10 | 1234 | |||
BC057446.1 | - | 10 | 1253 | 3155760 |