Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021535.4 | NP_067510.3 | 3 | 450 | ||
GenBank mRNA | AB044414.1 | - | 3 | 379 | ||
AK003493.1 | - | 3 | 437 | |||
AK011140.1 | - | 3 | 449 | |||
AK019174.1 | - | 3 | 409 | |||
AK145982.1 | - | 3 | 449 | |||
AK159635.1 | - | 3 | 437 | |||
AK168813.1 | - | 3 | 444 | |||
BC020034.1 | - | 3 | 392 | |||
BC057446.1 | - | 3 | 411 | 3155760 |